Merck
  • Utility of cardiac magnetic resonance (CMR) in the evaluation of right ventricular (RV) involvement in patients with myocardial infarction (MI).
Utility of cardiac magnetic resonance (CMR) in the evaluation of right ventricular (RV) involvement in patients with myocardial infarction (MI).
La Radiologia medica (2013-12-18)
Nicola Galea, Marco Francone, Iacopo Carbone, David Cannata, Francesco Vullo, Roberto Galea, Luciano Agati, Francesco Fedele, Carlo Catalano
ABSTRACT
The aim of this work was to compare the prevalence of right ventricle involvement in a population of patients with myocardial infarction as detected by cardiac magnetic resonance (CMR), clinical presentation, electrocardiographic (ECG) and echocardiographic criteria. A total of 97 consecutive patients, admitted to our institution for acute myocardial infarction, underwent a standard CMR examination within 5 days after the event. The presence of myocardial oedema and late enhancement of the right ventricle were compared to infarct location (anterior vs. inferior), clinical data, ECG, echocardiography results and other CMR findings. The results were analysed statistically using the Student's t test for independent samples and the K statistic. Among the 97 patients included in the study, a diagnosis of right ventricular infarction was established in 12, 14 and 24 cases on the basis of the clinical data, the ECG and echocardiography, respectively. CMR demonstrated myocardial oedema and late enhancement of the right ventricle in 48 and 32 cases, respectively. The right ventricle was involved in 46 % of patients with inferior myocardial infarction (15/32) and in 30 % with anterior myocardial infarction (17/56), correlating to a worsening of both right and left ventricular performance (p = 0.001-0.05). The right ventricle is frequently involved in myocardial infarction, correlating to a worse functional impairment of both ventricles and a worse prognosis. This finding, which is often underestimated by traditional cardiological tests, is well revealed by CMR, with potential clinical and therapeutic impact.
Sigma-Aldrich
N-Methyl-D-glucamine, ReagentPlus®, ≥99.0% (T)
Sigma-Aldrich
N-Methyl-D-glucamine, 99.0-100.5% (titration)
Supelco
Meglumine, Pharmaceutical Secondary Standard; Certified Reference Material
Meglumine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Meglumine, 99.0-100.5% dry basis, meets USP testing specifications