  • Search Results
  • The DNA damage response induces antigen presenting cell-like functions in fibroblasts.
The DNA damage response induces antigen presenting cell-like functions in fibroblasts.
European journal of immunology (2014-01-01)
Melissa Li Fang Tang, Muznah Khatoo Nazar Khan, John Ludovic Croxford, Kar Wai Tan, Veronique Angeli, Stephan Gasser
ABSTRACT
The DNA damage response (DDR) alerts the immune system to the danger posed by DNA damage through the induction of damage-associated molecular pattern molecules, chemokines, and ligands for activating immune receptors such as lymphocyte function-associated antigen 1 (LFA-1), NKG2D, and DNAX accessory molecule 1 (DNAM-1). Here we provide evidence that OVA(257-264) -pulsed fibroblasts gain the ability to activate naïve OT-I CD8(+) T cells in response to DNA damage. The ability of fibroblasts to activate OT-I CD8(+) T cells depended on the upregulation of ICAM-1 on fibroblasts and DNAM-1 expression of CD8(+) T cells. OVA(257-264) -pulsed fibroblasts were able to induce a protective T-cell response against B16-OVA cells in a DDR-dependent manner. Hence, the DDR may alert the immune system to the presence of potentially dangerous cells by upregulating the expression of ligands that can induce the activation of innate and adaptive immune cells.
Sigma-Aldrich
Cytosine β-D-arabinofuranoside, crystalline, ≥90% (HPLC)
Supelco
Albumin from chicken egg white, For use as a marker in SDS-PAGE
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥90% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from chicken egg white, powder, 62-88% (agarose gel electrophoresis)
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Cytosine β-D-arabinofuranoside hydrochloride, crystalline
Cytarabine, European Pharmacopoeia (EP) Reference Standard