Merck
  • A mechanical model of the cornea considering the crimping morphology of collagen fibrils.
A mechanical model of the cornea considering the crimping morphology of collagen fibrils.
Investigative ophthalmology & visual science (2014-04-03)
Xiaoyu Liu, Lizhen Wang, Jing Ji, Wei Yao, Wei Wei, Jie Fan, Shailesh Joshi, Deyu Li, Yubo Fan
ABSTRACT
To develop a mechanical model with which to investigate the relationship between the crimping morphology of collagen fibrils and the nonlinear mechanical behavior of the cornea. Uniaxial tensile experiments were performed with corneal strips to test their mechanical behavior. A constitutive model was constructed based on the Gaussian-distributed morphology of crimped collagen fibrils. The parameters that represent the micro characteristics of collagen fibrils were determined by fitting the experimental data to the constitutive model. Transmission electron microscopy (TEM) was used to visualize the crimping morphology of collagen fibrils in the stroma. A quantitative analysis of fibril crimping degrees in the TEM images was conducted to test the parameters predicted by the constitutive model. The parameters were derived using a fitting method that included the expectation for the distribution of fibril crimping degrees, μ = 1.063; the standard deviation, σ = 0.0781; the elastic modulus of collagen fibrils, E = 52.74 MPa; and the fibril ultimate strain, εb = 0.1957. TEM images showed a variation of the fibril crimping morphology when the cornea was subjected to different tensile loads. A good agreement was found between the parameters derived by the constitutive model and the data quantified from the TEM images. The nonlinear mechanical behavior of the cornea is closely correlated with the crimping morphology of collagen fibrils. The findings are expected to guide further research of corneal pathologies related to the abnormal microstructure of collagen fibrils.
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder