Merck
  • Efficacy of a novel topical combination of fipronil, (S)-methoprene, eprinomectin and praziquantel against larval and adult stages of the cat lungworm, Aelurostrongylus abstrusus.
Efficacy of a novel topical combination of fipronil, (S)-methoprene, eprinomectin and praziquantel against larval and adult stages of the cat lungworm, Aelurostrongylus abstrusus.
Veterinary parasitology (2014-04-08)
Martin Knaus, S Theodore Chester, Joseph Rosentel, Axel Kühnert, Steffen Rehbein
ABSTRACT
The efficacy of a novel topical combination of fipronil 8.3% w/v, (S)-methoprene 10% w/v, eprinomectin 0.4% w/v, and praziquantel 8.3% w/v (BROADLINE(®),(1) Merial) against larval and adult Aelurostrongylus abstrusus lungworms in cats was assessed in a controlled laboratory study. The study included 48 purpose-bred, short-haired cats which were each inoculated with 225 infective A. abstrusus larvae. The cats were formed into eight blocks based on pre-treatment bodyweight and were then, within each block, randomly allocated to one of six treatment groups: untreated control; treated once when A. abstrusus were expected to be third-stage larvae (4 days post inoculation [dpi]), fourth-stage larvae (7 dpi), immature adults (14 dpi) or adult nematodes (32 dpi), or treated twice, once when A. abstrusus were expected to be third-stage larval and once again when A. abstrusus were expected to be adult nematodes (4 dpi+32 dpi). Cats weighing ≥ 0.8-2.5 kg received one 0.3 mL applicator and cats weighing >2.5-7.5 kg received one 0.9 mL applicator. For determination of the efficacy of treatments, lungworm larval counts were established on faecal samples collected from all cats 32, 39, 46, 53 and 60 dpi. At each occasion from 46 dpi on, cats treated with fipronil, (S)-methoprene, eprinomectin and praziquantel had significantly lower A. abstrusus larval counts than the untreated controls with percentage reductions of 91.6% (cats treated 14dpi; P=0.012), ≥ 98.9% (cats treated either 4 dpi, 7 dpi or 32 dpi; P<0.001) or >99.9% (cats treated 4 dpi+32 dpi; P<0.001) at 60 dpi. Thus, the novel topical combination of fipronil, (S)-methoprene, eprinomectin and praziquantel was highly effective in the prevention and treatment of A. abstrusus lungworm infection in cats.
Product Description
Supelco
Eprinomectin, PESTANAL®, analytical standard, mixture of isomers
Supelco
Fipronil, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Fipronil, PESTANAL®, analytical standard
Praziquantel for system suitability, European Pharmacopoeia (EP) Reference Standard
Ivermectin, European Pharmacopoeia (EP) Reference Standard
Praziquantel, European Pharmacopoeia (EP) Reference Standard
Supelco
Praziquantel, VETRANAL®, analytical standard
Sigma-Aldrich
Ivermectin
Supelco
Methoprene, PESTANAL®, analytical standard
Supelco
Praziquantel, Pharmaceutical Secondary Standard; Certified Reference Material