Merck
  • Induction of osteogenic differentiation by nanostructured alumina surfaces.
Induction of osteogenic differentiation by nanostructured alumina surfaces.
Journal of biomedical nanotechnology (2014-04-17)
Wolfgang Metzger, Benedikt Schwab, Marina M Miro, Sibylle Grad, Angharad Simpson, Michael Veith, Gunther Wennemuth, Vladimir Zaporojtchenko, Sophie Verrier, Jessica S Hayes, Monika Bubel, Tim Pohlemann, Martin Oberringer, Cenk Aktas
ABSTRACT
Permanent orthopedic implants are becoming increasingly important due to the demographic development. Their optimal osseointegration is key in obtaining good secondary stability. For anchorage dependent cells, topographic features of a surface play an essential role for cell adhesion, proliferation, differentiation and biomineralization. We studied the topographical effect of nanostructured alumina surfaces prepared by chemical vapor deposition on osteogenic differentiation and growth of human osteoblasts. Chemical vapor deposition of the single source precursor (tBuOAIH2)2 led to synthesis of one dimensional alumina nanostructures of high purity with a controlled stoichiometry. We fabricated different topographic features by altering the distribution density of deposited one dimensional nanostructures. Although the topography differed, all surfaces exhibited identical surface chemistry, which is the key requirement for systematically studying the effect of the topography on cells. Forty-eight hours after seeding, cell density and cell area were not affected by the nanotopography, whereas metabolic activity was reduced and formation of actin-fibres and focal adhesions was impaired compared to the uncoated control. Induction of osteogenic differentiation was demonstrated via up-regulation of alkaline phosphatase, bone sialoprotein, osteopontin and Runx2 at the mRNA level, demonstrating the potential of nanostructured surfaces to improve the osseointegration of permanent implants.
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Supelco
Aluminum oxide, for the determination of hydrocarbons
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Aluminum oxide, standard grade, Brockmann I, activated, basic
Sigma-Aldrich
Aluminum oxide, Brockmann I, standard grade, activated, acidic
Sigma-Aldrich
Aluminum oxide, pore size 58 Å, ~150 mesh
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, 100-200 mesh
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
Aluminum oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum oxide, Corundum, α-phase, -100 mesh
Sigma-Aldrich
Aluminum oxide, activated, Brockmann I, standard grade, neutral
Sigma-Aldrich
Aluminum oxide, Type WN-6, Neutral, Activity Grade Super I
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I