Merck
  • Controlled release by using poly (methacrylic acid-g-ethylene glycol) hydrogels
Controlled release by using poly (methacrylic acid-g-ethylene glycol) hydrogels
Journal of Controlled Release : Official Journal of the Controlled Release Society (1991)
Peppas, N. A., & Klier, J.