Merck
  • Lovastatin extended release: a review of its use in the management of hypercholesterolaemia.
Lovastatin extended release: a review of its use in the management of hypercholesterolaemia.
Drugs (2003-03-27)
Monique P Curran, Karen L Goa
ABSTRACT
Lovastatin extended release (ER) provides a new form of delivery for lovastatin, an HMG-CoA reductase inhibitor. Lovastatin ER delivers the drug in a more sustained fashion, as shown by a smoother plasma concentration-time profile, a lower maximum plasma concentration and a prolonged half-life compared with that of lovastatin immediate release (IR). At dosages of 10-60 mg/day, lovastatin ER significantly reduced levels of total cholesterol, low density lipoprotein (LDL)-cholesterol and triglycerides, and increased levels of high density lipoprotein-cholesterol, in patients with primary hypercholesterolaemia in a randomised, double-blind study of 12 weeks' duration. These effects were maintained in a 6-month extension study in which patients received lovastatin 40 or 60 mg/day. In a randomised 4-week study in 24 patients with primary hypercholesterolaemia, the reduction in plasma LDL-cholesterol levels was significantly greater with lovastatin ER 40 mg/day than with the IR formulation administered at the same dosage. Lovastatin ER was well tolerated in all studies and adverse events were usually mild to moderate and transient. The tolerability profile of lovastatin ER was similar to that of lovastatin IR. There were no reports of clinically relevant elevations in liver transaminases or creatine phosphokinase attributed to the drug in recipients of lovastatin ER. The ER formulation of lovastatin provides smooth and sustained delivery of this established and well-tolerated agent over the dosage interval, significantly reducing LDL-cholesterol in patients with primary hypercholesterolaemia. If, as expected, the beneficial changes in lipid levels are maintained during long-term treatment and further clinical experience confirms the greater efficacy of the lovastatin ER formulation than the IR formulation, then lovastatin ER is likely to supplant lovastatin IR and provide a useful option in the management of patients with dyslipidaemia and prevention of coronary heart disease.
Product Description
USP
Lovastatin, United States Pharmacopeia (USP) Reference Standard
Supelco
Lovastatin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Mevinolin from Aspergillus sp., ≥98% (HPLC)
Lovastatin, European Pharmacopoeia (EP) Reference Standard
Lovastatin for peak identification, European Pharmacopoeia (EP) Reference Standard