- EFFECT OF LYSERGIC ACID DIETHYLAMIDE ON POTENTIALS EVOKED IN THE SPECIFIC SENSORY PATHWAYS.
EFFECT OF LYSERGIC ACID DIETHYLAMIDE ON POTENTIALS EVOKED IN THE SPECIFIC SENSORY PATHWAYS.
British medical bulletin (1965-01-01)
B J KEY
PMID14292497
Product Description
Supelco
LSD solution, 25 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
LSD solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®