Merck
  • The role of Triclosan in dentifrice formulations, with particular reference to a new 0.3% Triclosan calcium carbonate-based system.
The role of Triclosan in dentifrice formulations, with particular reference to a new 0.3% Triclosan calcium carbonate-based system.
International dental journal (2004-10-29)
M G Brading, V J Cromwell, A K Green, S DeBrabander, T Beasley, P D Marsh
ABSTRACT
To discuss the role of Triclosan in dentifrice systems and demonstrate the enhanced efficacy of Triclosan in calcium carbonate-based systems when the level of the antimicrobial agent is raised from 0.2% to 0.3%. Triclosan is the most commonly used antimicrobial agent in oral care products, being compatible with a wide range of ingredients found in toothpaste formulations, whilst having no negative sensory features (e.g. taste, staining) that are associated with some other antimicrobial/anti-plaque agents. Triclosan is a broad spectrum antimicrobial agent, with additional anti-metabolic and anti-inflammatory properties. When delivered to the mouth in oral care products, Triclosan can selectively inhibit Gram negative anaerobic bacteria implicated in gingivitis and periodontal diseases, while leaving species associated with oral health relatively unaffected. Worldwide, attempts have been made to boost delivery/activity of Triclosan, either by use of copolymers or by combination with other agents such as zinc citrate. However, Triclosan has also been shown to maintain clinical efficacy against plaque and gingivitis when present as the sole antimicrobial in toothpaste formulations.
Product Description
USP
Calcium Carbonate (AS), United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Triclosan, 97.0-103.0% (active substance, GC)
Supelco
Calcium carbonate, reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Calcium carbonate, ACS reagent, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Supelco
Triclosan, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Calcium carbonate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder
Sigma-Aldrich
Calcium carbonate, ReagentPlus®
Sigma-Aldrich
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)