Merck
  • Concentrations of toluene in the body killed by an injury to the head shortly after ingesting thinner.
Concentrations of toluene in the body killed by an injury to the head shortly after ingesting thinner.
Forensic science international (2004-11-16)
Yukihito Yajima, Masato Funayama, Hisae Niitsu, Masayuki Nata, Yoshimasa Kanawaku, Jun Sakai, Yasuhiro Aoki
ABSTRACT
An autopsy was conducted on a male showing leather-like skin damage, revealing the cause of death to be an injury to the head. Thinner was found scattered around the scene of death, and stomach and intestine contents smelled strongly of solvent. Toxicological analysis was conducted to determine whether or not the solvent was of a lethal level. Using gas chromatography, peaks of toluene, xylene, and ethylbenzene were detected in the blood and gastric contents. No toluene was detected in the urine, and therefore it was concluded that the decedent died of a severe head injury shortly after solvent ingestion. In the literature, toluene concentrations in blood and lung samples were determined as both fatal and non-fatal but clear differences in the fatality of toluene in solid organ samples, namely, the brain, liver and kidneys were shown. The brain is especially useful in postmortem analysis. In this case, the concentration of toluene in the brain was 20.0 microl/g, which was considered as a non-lethal level.
Product Description
Supelco
Toluene solution, certified reference material, 5000 μg/mL in methanol
Supelco
Toluene solution, certified reference material, TraceCERT®, 200 μg/mL in methanol, ampule of 1 mL
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Toluene, analytical standard
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%