Merck
  • Epidemiology, toxicokinetics, and health effects of methyl tert-butyl ether (MTBE).
Epidemiology, toxicokinetics, and health effects of methyl tert-butyl ether (MTBE).
Journal of medical toxicology : official journal of the American College of Medical Toxicology (2008-06-24)
Scott Phillips, Robert B Palmer, Aaron Brody
ABSTRACT
This paper reviews the published information assessing the kinetics and potential for adverse health effects related to exposure to the fuel oxygenate, methyl tert-butyl ether (MTBE). Data were obtained from previously published reports, using human data where possible. If human data were not available, animal studies were cited. The kinetic profile of MTBE in humans is similar for ingestion and inhalation. The concentrations of MTBE to which the general public is expected to be exposed are orders of magnitude below concentrations that have caused adverse health effects in animals. Controlled human studies have not replicated early epidemiology studies that suggested, but did not confirm, a possible association between MTBE exposure and nonspecific health complaints.
Sigma-Aldrich
tert-Butyl methyl ether, suitable for HPLC, ≥99.8%
Sigma-Aldrich
tert-Butyl methyl ether, ACS reagent, ≥99.0%
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, ≥98%
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Supelco
tert-Butyl methyl ether, analytical standard