Merck
[Bullous scabies responding to ivermectin therapy].
Actas dermo-sifiliograficas (2010-01-30)
L Galvany Rossell, M Salleras Redonnet, P Umbert Millet
ABSTRACT
Bullous scabies is a rare disease that is usually diagnosed in elderly patients. The clinical, histological, and immunological findings are identical to bullous pemphigoid. In a review of the literature, we found reports of 24 cases. We present a new case of bullous scabies in a 72-year-old man. The lesions responded to treatment with oral ivermectin.
Product Description
Ivermectin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ivermectin