Merck
[Progresses in research of hemolysis induced by puerarin injection].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2011-08-16)
Qin Wang, Xiaoyu Xu
ABSTRACT
Puerarin injection has been widely used for the treatment of cardiac/cerebral blood vascular diseases. The clinical statistical investigation had confirmed that puerarin injection can induce acute intravascular hemolysis, which effected the use of puerarin injection seriously. The review was focus on the research progress of the hemolysis of puerarin injection including the work in laboratory and clinic. The potential hemolysis mechanism and allergen of the puerarin injection were summarized and discussed.
Sigma-Aldrich
Puerarin, ≥98.0% (HPLC)
Supelco
Puerarin, analytical standard