Merck
  • Levetiracetam may worsen myoclonus in patients with juvenile myoclonic epilepsy: case reports.
Levetiracetam may worsen myoclonus in patients with juvenile myoclonic epilepsy: case reports.
Clinical neuropharmacology (2012-07-19)
Fawzi A Babtain
ABSTRACT
Levetiracetam was approved for generalized and partial epilepsy in pediatric and adult population. It is also an effective antimyoclonus, but the evidence only supports its use as an adjunctive agent along with other antiepileptic drugs, such as sodium valproate, and it is commonly used in cases with juvenile myoclonic epilepsy. We report here 2 cases with juvenile myoclonic epilepsy who were switched from sodium valproate to levetiracetam to avoid the cosmetic or future teratogenic effect, but this switch was associated with exaggerated myoclonus despite escalating the dose of levetiracetam but resolved completely after reintroducing sodium valproate.
Product Description
USP
Levetiracetam, United States Pharmacopeia (USP) Reference Standard
Supelco
Levetiracetam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Levetiracetam, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Levetiracetam, ≥98% (HPLC)
Supelco
Levetiracetam, Pharmaceutical Secondary Standard; Certified Reference Material