Merck
Metabolic and endocrine effects of valproic acid chronic treatment.
Epilepsy research (2013-10-01)
Vincenzo Belcastro, Claudia D'Egidio, Pasquale Striano, Alberto Verrotti
ABSTRACT
Treatment of epileptic patients with valproic acid (VPA) may be associated with substantial weight changes that may increase morbidity and impair adherence to the treatment regimen. VPA-induced weight gain seems to be associated with many metabolic disturbances; the most frequent are hyperinsulinemia and insulin resistance, hyperleptinemia and leptin resistance. Patients who gain weight during VPA therapy can develop dyslipidemia and metabolic syndrome that are associated with long-term vascular complications such as hypertension and atherosclerosis. Moreover, an elevation in the levels of uric acid and homocysteine, together with oxidative stress, may contribute to atherosclerotic risk in patients under long-term therapy with VPA. The aim of this review is to discuss the metabolic and endocrine effects of VPA chronic treatment in patients with epilepsy.
Product Description
Supelco
Valproic acid solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Valproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propylpentanoic acid
USP
Valproic acid, United States Pharmacopeia (USP) Reference Standard
Valproic acid, European Pharmacopoeia (EP) Reference Standard