- Luminescent gold nanoparticles: a new class of nanoprobes for biomedical imaging.
Luminescent gold nanoparticles: a new class of nanoprobes for biomedical imaging.
Experimental biology and medicine (Maywood, N.J.) (2013-10-11)
Chen Zhou, Shengyang Yang, Jinbin Liu, Mengxiao Yu, Jie Zheng
PMID24108449
ABSTRACT
Our fundamental understanding of cell biology and early diagnosis of human diseases have been greatly benefited from the development of fluorescent probes. Over the past decade, luminescent gold nanoparticles (AuNPs) with diverse structural parameters including particle size, surface ligands, valence state and grain size have been synthesized and have begun to emerge as a new class of fluorescent probes for bioimaging because of their great biocompatibility, robust photophysical properties and tunable emissions from the visible range to the near infrared region. In this minireview, we summarize the recent progress in applications of different-sized luminescent AuNPs as imaging probes for both in vitro and in vivo levels.
Product Description
Gold, rod, 6mm, diameter 6.0mm, as drawn, 99.95%
Gold, tube, 50mm, outside diameter 8mm, inside diameter 7.8mm, wall thickness 0.1mm, as drawn, 99.95%
Gold, rod, 50mm, diameter 3.0mm, as drawn, 99.95%
Gold, tube, 200mm, outside diameter 2.0mm, inside diameter 1.8mm, wall thickness 0.1mm, as drawn, 99.95%
Gold, rod, 25mm, diameter 4.0mm, as drawn, 99.95%
Gold, tube, 100mm, outside diameter 1.60mm, inside diameter 0.6mm, wall thickness 0.5mm, as drawn, 99.95%
Gold, tube, 200mm, outside diameter 1.0mm, inside diameter 0.5mm, wall thickness 0.25mm, as drawn, 99.95%
Gold, tube, 100mm, outside diameter 10.0mm, inside diameter 9.7mm, wall thickness 0.15mm, as drawn, 99.95%
Gold, insulated wire, 0.5m, conductor diameter 0.125mm, insulation thickness 0.016mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Gold, insulated wire, 0.5m, conductor diameter 0.125mm, insulation thickness 0.014mm, polyester insulation, 99.99%
Gold, insulated wire, 5m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyester insulation, 99.99%
Gold, microleaf disks, 10mm, thinness 0.5μm, specific density 966μg/cm2, removable support, 99.99+%
Gold, insulated wire, 0.5m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyester insulation, 99.99%
Gold, insulated wire, 1m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyester insulation, 99.99%
Gold, microfoil, disks, 10mm, thinness 0.01μm, specific density 20.7μg/cm2, permanent mylar 3.5μm support, 99.99+%
Gold, insulated wire, 1m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyimide insulation, 99.99%
Gold, insulated wire, 2m, conductor diameter 0.075mm, insulation thickness 0.012mm, PTFE (polytetrafluoroethylene) insulation, 99.99%