Merck
  • Chitosan/hyaluronic acid polyelectrolyte complex hydrogels in the management of burn wounds.
Chitosan/hyaluronic acid polyelectrolyte complex hydrogels in the management of burn wounds.
Revista medico-chirurgicala a Societatii de Medici si Naturalisti din Iasi (2013-12-18)
Cornelia Vasile, D Pieptu, Raluca Petronela Dumitriu, Andreea Pânzariu, Lenuţa Profire
ABSTRACT
This paper is a short review on the state-of-the-art of the use of polyelectrolyte complexes containing polysaccharides for wound and burn healing and treatment using the chitosan/hyaluronic acid polyelectrolyte complex (PEC) hydrogel. PEC is suitable for wound healing because a wet treatment can be realized and both components of PEC contribute by their properties to the enhanced antimicrobial activity, promote wound healing and prevent wound damage during treatment.
Sigma-Aldrich
Chitosan from shrimp shells, low-viscous
Sigma-Aldrich
Chitosan, from shrimp shells, ≥75% (deacetylated)
Sigma-Aldrich
Chitosan, high molecular weight
Sigma-Aldrich
Chitosan, low molecular weight
Sigma-Aldrich
Chitosan, medium molecular weight
Sigma-Aldrich
Chitosan, from shrimp shells, practical grade