Merck
  • Calcium oxalate monohydrate renal calculi. Formation and development mechanism.
Calcium oxalate monohydrate renal calculi. Formation and development mechanism.
Advances in colloid and interface science (1995-08-31)
O Söhnel, F Grases
ABSTRACT
Available information relevant to stone genesis on both calcium oxalate monohydrate crystallization and fine inner structure of papillary calculi is reviewed. Integration of attained facts facilitated formulating a feasible mechanism of papillary calculi formation and development. Medical implications of this mechanism are assessed.
Product Description
Calcium oxalate monohydrate, European Pharmacopoeia (EP) Reference Standard