Merck
  • Assessment of structurally related chemicals: toxicity and ecotoxicity of acrylic acid and acrylic acid alkyl esters (acrylates), methacrylic acid and methacrylic acid alkyl esters (methacrylates).
Assessment of structurally related chemicals: toxicity and ecotoxicity of acrylic acid and acrylic acid alkyl esters (acrylates), methacrylic acid and methacrylic acid alkyl esters (methacrylates).
Chemosphere (1995-07-01)
H Greim, J Ahlers, R Bias, B Broecker, H Hollander, H P Gelbke, S Jacobi, H J Klimisch, I Mangelsdorf, W Mayr
ABSTRACT
BUA compiled the available data on toxicity and ecotoxicity for several acrylic and methacrylic acid esters and their corresponding acids. A comparison of these data revealed a qualitative similarity in the toxicological and ecotoxicological properties of the compounds considered. The data indicate that methacrylates are less reactive than the corresponding acrylates.
Sigma-Aldrich
Acrylic acid, anhydrous, contains 200 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Methacrylic acid, contains 250 ppm MEHQ as inhibitor, 99%