Merck
  • The anaerobic degradation of L-serine and L-threonine in enterobacteria: networks of pathways and regulatory signals.
The anaerobic degradation of L-serine and L-threonine in enterobacteria: networks of pathways and regulatory signals.
Archives of microbiology (1998-12-31)
G Sawers
ABSTRACT
The mechanisms controlling the biosynthesis and degradation of l-serine and l-threonine are remarkably complex. Their metabolism forms a network of pathways linking several amino acids, central primary metabolites such as pyruvate, oxaloacetate and 3-phosphoglycerate, and C1 metabolism. Studies on the degradation of these amino acids in Escherichia coli have revealed the involvement of fascinating enzymes that utilise quite diverse catalytic mechanisms. Moreover, it is emerging that both environmental and metabolic signals have a major impact in controlling enzyme synthesis This is exemplified by the anaerobically regulated tdc operon, which encodes a metabolic pathway for the degradation of serine and threonine. Studies on this pathway are beginning to provide insights into how an organism adapts its genetic makeup to meet the physiological demands of the cell.
Product Description
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Serine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
SAFC
L-Threonine
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
L-Threonine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Serine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Serine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Serine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Serine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Serine, BioUltra, ≥99.5% (NT)