Merck
  • Triple therapy with methotrexate, sulfasalazine, and hydroxychloroquine in patients with rheumatoid arthritis.
Triple therapy with methotrexate, sulfasalazine, and hydroxychloroquine in patients with rheumatoid arthritis.
Rheumatic diseases clinics of North America (1998-08-26)
J R O'Dell
ABSTRACT
Combinations of DMARDs are currently used to treat rheumatoid arthritis by almost all rheumatologists. This article reviews the published data on the triple combination of methotrexate, sulfasalazine, and hydroxychloroquine, discusses caveats for clinical use, compares efficacy of different combinations, and speculates on future combination therapies.
Sigma-Aldrich
Sulfasalazine, 97.0-101.5%