Merck
  • Anacardic acid derivatives as inhibitors of glyceraldehyde-3-phosphate dehydrogenase from Trypanosoma cruzi.
Anacardic acid derivatives as inhibitors of glyceraldehyde-3-phosphate dehydrogenase from Trypanosoma cruzi.
Bioorganic & medicinal chemistry (2008-09-16)
Junia M Pereira, Richele P Severino, Paulo C Vieira, João B Fernandes, M Fátima G F da Silva, Aderson Zottis, Adriano D Andricopulo, Glaucius Oliva, Arlene G Corrêa
ABSTRACT
Chagas' disease, a parasitic infection caused by the flagellate protozoan Trypanosoma cruzi, is a major public health problem affecting millions of individuals in Latin America. On the basis of the essential role in the life cycle of T. cruzi, the glycolytic enzyme glyceraldehyde-3-phosphate dehydrogenase (GAPDH) has been considered an attractive target for the development of novel antitrypanosomatid agents. In the present work, we describe the inhibitory effects of a small library of natural and synthetic anacardic acid derivatives against the target enzyme. The most potent inhibitors, 6-n-pentadecyl- and 6-n-dodecylsalicilic acids, have IC(50) values of 28 and 55 microM, respectively. The inhibition was not reversed or prevented by the addition of Triton X-100, indicating that aggregate-based inhibition did not occur. In addition, detailed mechanistic characterization of the effects of these compounds on the T. cruzi GAPDH-catalyzed reaction showed clear noncompetitive inhibition with respect to both substrate and cofactor.