Merck
  • N-Methylsuccinimide in plasma and urine as a biomarker of exposure to N-methyl-2-pyrrolidone.
N-Methylsuccinimide in plasma and urine as a biomarker of exposure to N-methyl-2-pyrrolidone.
International Archives of Occupational and Environmental Health (2004)
Jonsson BAG and ?kesson B.