- A sugar-quinoline fluorescent chemosensor for selective detection of Hg2+ ion in natural water.
A sugar-quinoline fluorescent chemosensor for selective detection of Hg2+ ion in natural water.
Chemical communications (Cambridge, England) (2006-10-24)
Shengju Ou, Zhihua Lin, Chunying Duan, Haitao Zhang, Zhiping Bai
PMID17057854
ABSTRACT
A selective and sensitive fluorescent sensor for detection of Hg2+ in natural water was achieved by incorporating the well-known fluorophore quinoline group and a water-soluble D-glucosamine group within one molecule.