Merck
  • Nonmetal haptens induce ATP release from keratinocytes through opening of pannexin hemichannels by reactive oxygen species.
Nonmetal haptens induce ATP release from keratinocytes through opening of pannexin hemichannels by reactive oxygen species.
The Journal of investigative dermatology (2014-02-18)
Kaoru Onami, Yutaka Kimura, Yumiko Ito, Takeshi Yamauchi, Kenshi Yamasaki, Setsuya Aiba
ABSTRACT
Although extracellular adenosine 5'-triphosphate (eATP) has a crucial role in the sensitization phase of contact hypersensitivity (CHS), the mechanism by which hapten causes keratinocyte cell death and ATP release is unknown. We examined the time course of cell death, reactive oxygen species (ROS) production, and ATP release in HaCaT cells and in normal human keratinocytes after exposure to nonmetal haptens, NiCl2, or irritants. Both haptens and irritants caused cell death of keratinocytes but with different time courses. N-acetylcysteine (NAC) significantly reduced only nonmetal hapten-induced cell death as assessed by propidium iodide exclusion. We examined the effects of antioxidants and pannexin (Panx) inhibitors on cell death, ROS production, and ATP release by chemical-treated HaCaT cells. Nonmetal hapten-induced cell death, but not NiCl2- or irritant-related cell death, was dependent on reactivity to thiol residues in the cells. NAC reduced cell death and ATP release, whereas antioxidants and Panx inhibitors did not inhibit cell death but significantly attenuated ATP release. Panx1 small interfering RNA (siRNA) also suppressed ATP release from hapten-exposed HaCaT cells. Intraperitoneal injection of a Panx1 inhibitor attenuated murine CHS. These findings suggest that nonmetal hapten reactivity to thiol residues causes membrane disruption of keratinocytes and ROS production that leads to ATP release through opening of Panx hemichannels.
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, ≥99%
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Supelco
L-Cystine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, 97%
SAFC
L-Cystine
Sigma-Aldrich
Nickel(II) chloride hydrate, 99.95% trace metals basis
Sigma-Aldrich
Nickel(II) chloride hexahydrate, 99.999% trace metals basis
Sigma-Aldrich
L-Cystine, ≥98% (TLC), crystalline
Sigma-Aldrich
L-Cystine, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Nickel(II) chloride hexahydrate, BioReagent, suitable for cell culture
Sigma-Aldrich
Nickel(II) chloride hexahydrate, 99.9% trace metals basis
Sigma-Aldrich
L-Cystine, Wacker Chemie AG, ≥98.5%
Sigma-Aldrich
L-Cystine, ≥99.7% (TLC)
Nickel, foil, not light tested, 25x25mm, thickness 0.001mm, 99.95%
Sigma-Aldrich
Nickel(II) chloride hexahydrate, puriss. p.a., ≥98%
Sigma-Aldrich
Nickel(II) chloride hexahydrate, ReagentPlus®
Nickel, foil, light tested, 25x25mm, thickness 0.009mm, 99.95%
Nickel, foil, not light tested, 25x25mm, thickness 0.0005mm, temporary acrylic support, 99.95%
Nickel, foil, light tested, 100x100mm, thickness 0.007mm, 99.95%
Nickel, foil, not light tested, 100x100mm, thickness 0.004mm, hard, 99%
Nickel, foil, 4mm disks, thickness 0.3mm, as rolled, 100%
Cystine, European Pharmacopoeia (EP) Reference Standard
Nickel, foil, 10mm disks, thickness 0.5mm, as rolled, 100%
Nickel, foil, 10mm disks, thickness 0.05mm, as rolled, 99.99+%
Nickel, rod, 200mm, diameter 30mm, 99%