Merck
  • Effects of environmental levels of cadmium, lead and mercury on human renal function evaluated by structural equation modeling.
Effects of environmental levels of cadmium, lead and mercury on human renal function evaluated by structural equation modeling.
Toxicology letters (2014-04-29)
Jerome P Trzeciakowski, Lesley Gardiner, Alan R Parrish
ABSTRACT
A relationship between exposure to heavy metals, including lead and cadmium, and renal dysfunction has long been suggested. However, modeling of the potential additive, or synergistic, impact of metals on renal dysfunction has proven to be challenging. In these studies, we used structural equation modeling (SEM), to investigate the relationship between heavy metal burden (serum and urine levels of lead, cadmium and mercury) and renal function using data from the NHANES database. We were able to generate a model with goodness of fit indices consistent with a well-fitting model. This model demonstrated that lead and cadmium had a negative relationship with renal function, while mercury did not contribute to renal dysfunction. Interestingly, a linear relationship between lead and loss of renal function was observed, while the maximal impact of cadmium occurred at or above serum cadmium levels of 0.8 μg/L. The interaction of lead and cadmium in loss of renal function was also observed in the model. These data highlight the use of SEM to model interaction between environmental contaminants and pathophysiology, which has important implications in mechanistic and regulatory toxicology.
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis
Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis
Lead, foil, not light tested, 25x25mm, thickness 0.0125mm, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, 8mm disks, thickness 0.01mm, 99.99+%
Lead, foil, 1m coil, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 50x50mm, thickness 6.0mm, as rolled, 99.99+%
Lead, foil, 10mm disks, thickness 0.04mm, 99.99+%
Lead, foil, 50x50mm, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 6mm disks, thickness 1.0mm, as rolled, 99.99+%
Lead, foil, 10mm disks, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 6mm disks, thickness 0.008mm, 99.99+%
Lead, foil, 25mm disks, thickness 0.03mm, 99.99+%
Lead, foil, 150x150mm, thickness 0.125mm, as rolled, 99.95%
Lead, foil, 10mm disks, thickness 0.007mm, 99.99+%
Lead, foil, 0.5m coil, thickness 0.016mm, as rolled, 96% (sn 1.5%, sb 2.5%)
Lead, foil, 6mm disks, thickness 0.125mm, as rolled, 99.95%
Lead, foil, 1m coil, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 5m coil, thickness 0.016mm, as rolled, 96% (sn 1.5%, sb 2.5%)
Lead, foil, 15mm disks, thickness 0.55mm, as rolled, 100%
Lead, foil, 6mm disks, thickness 0.009mm, 99.99+%
Lead, foil, 50mm disks, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 10mm disks, thickness 0.008mm, 99.99+%
Lead, foil, 15mm disks, thickness 0.006mm, 99.99+%
Lead, foil, 15mm disks, thickness 0.02mm, 99.99+%
Lead, foil, 15mm disks, thickness 0.5mm, as rolled, 99.99+%
Lead, foil, 25x25mm, thickness 0.55mm, as rolled, 99.999%
Lead, foil, 6mm disks, thickness 0.0125mm, 99.99+%
Lead, foil, 300x300mm, thickness 0.5mm, as rolled, 99.95%