Merck
  • 6-trifluoromethyl-2-thiouracil possesses anti-Toxoplasma gondii effect in vitro and in vivo with low hepatotoxicity.
6-trifluoromethyl-2-thiouracil possesses anti-Toxoplasma gondii effect in vitro and in vivo with low hepatotoxicity.
Experimental parasitology (2014-05-17)
Hwa-Jung Choi, Seung-Taek Yu, Kee-In Lee, Joong-Kwon Choi, Bo-Yoon Chang, Sung-Yeon Kim, Mi-Hwa Ko, Hyun-Ok Song, Hyun Park
ABSTRACT
Since pyrimethamine, the general therapeutic drug for toxoplasmosis, presents several adverse side effects, the need to develop and evaluate new drugs for the condition is critical. In this study, anti-Toxoplasma gondii activities of 3-[{2-((E)-furan-2-ylmethylene)hydrazinyl}methylene]-1,3-dihydroindol-2-one (ATT-5126) and 6-trifluoromethyl-2-thiouracil (KH-0562) were evaluated in vitro using a 3-(4,5-dimethylthiazol-2-yl)-5-(3-carboxymethoxyphenyl)-2-(4-sulfophenyl)-2H-tetrazolium (MTS) assay and in vivo by measuring amount of the tachyzoites in mice ascites. Biochemical parameters such as lipid peroxidation (LPO), glutathione (GSH), alanine aminotransferase (ALT), and aspartate aminotransferase (AST) were also evaluated in livers of mice at 4 days post-infection. As a result, the ATT-5126 and KH-0562 showed anti-T. gondii activity in vitro. Treatment of ATT-5126 or KH-0562 decreased the amount of tachyzoites in T. gondii infected ICR mice. The relative weight of liver and spleen increased by T. gondii infection were decreased by treatment of ATT-5126 or KH-0562. The levels of LPO, ALT and AST, which are biochemical parameters involved in liver injury, were also significantly recovered by treatment of ATT-5126 or KH-0562 (p<0.05). In particular, the recovered levels by KH-0562 were similar to those of pyrimethamine-treated group (p<0.05). However, the level of GSH, which is an antioxidant indicator, showed insignificant statistics. The results suggest that KH-0562 show anti-T. gondii activities in vitro and in vivo with low hepatotoxicity. Therefore, KH-0562 may be a useful candidate for treating T. gondii infection.
Product Description
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
USP
Pyrimethamine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, analytical standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Supelco
Pyrimethamine, VETRANAL®, analytical standard
Pyrimethamine, European Pharmacopoeia (EP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Thiouracil, ≥99%
Glutathione, European Pharmacopoeia (EP) Reference Standard
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material