Merck
  • Fracture prediction after discontinuation of 4 to 5 years of alendronate therapy: the FLEX study.
Fracture prediction after discontinuation of 4 to 5 years of alendronate therapy: the FLEX study.
JAMA internal medicine (2014-05-07)
Douglas C Bauer, Ann Schwartz, Lisa Palermo, Jane Cauley, Marc Hochberg, Art Santora, Steven R Cummings, Dennis M Black
ABSTRACT
Discontinuation of bisphosphonate therapy after 3 to 5 years is increasingly considered, but methods to monitor fracture risk after discontinuation have not been established. To test methods of predicting fracture risk among women who have discontinued alendronate therapy after 4 to 5 years. The prospective Fracture Intervention Trial Long-term Extension (FLEX) study randomized postmenopausal women aged 61 to 86 years previously treated with 4 to 5 years of alendronate therapy to 5 more years of alendronate or placebo from 1998 through 2003; the present analysis includes only the placebo group. Hip and spine dual-energy x-ray absorptiometry (DXA) were measured when placebo was begun (FLEX baseline) and after 1 to 3 years of follow-up. Two biochemical markers of bone turnover, urinary type 1 collagen cross-linked N-telopeptide (NTX) and serum bone-specific alkaline phosphatase (BAP), were measured at FLEX baseline and after 1 and 3 years. Symptomatic spine and nonspine fractures occurring after the follow-up measurement of DXA or bone turnover. During 5 years of placebo, 94 of 437 women (22%) experienced 1 or more symptomatic fractures; 82 had fractures after 1 year. One-year changes in hip DXA, NTX, and BAP were not related to subsequent fracture risk, but older age and lower hip DXA at time of discontinuation were significantly related to increased fracture risk (lowest tertile of baseline femoral neck DXA vs other 2 tertiles relative hazard ratio, 2.17 [95% CI, 1.38-3.41]; total hip DXA relative hazard ratio, 1.87 [95% CI, 1.20-2.92]). Among postmenopausal women who discontinue alendronate therapy after 4 to 5 years, age and hip BMD at discontinuation predict clinical fractures during the subsequent 5 years. Follow-up measurements of DXA 1 year after discontinuation and of BAP or NTX 1 to 2 years after discontinuation are not associated with fracture risk and cannot be recommended. clinicaltrials.gov Identifier: NCT00398931.
Sigma-Aldrich
Phosphatase, Alkaline bovine, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein
USP
Alendronate sodium, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phosphatase, Alkaline shrimp, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host
Sigma-Aldrich
Alendronate sodium trihydrate, ≥97% (NMR), powder
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from calf intestinal mucosa, suitable for enzyme immunoassay, solution (clear, colorless), ~2500 U/mg protein (~10 mg/ml)
Sigma-Aldrich
Phosphatase, Alkaline from porcine kidney, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, ≥5,700 DEA units/mg protein