Merck
  • In vitro model to study the biomaterial-dependent reaction of macrophages in an inflammatory environment.
In vitro model to study the biomaterial-dependent reaction of macrophages in an inflammatory environment.
The British journal of surgery (2014-05-20)
N Grotenhuis, H F E Vd Toom, N Kops, Y Bayon, E B Deerenberg, I M Mulder, G J V M van Osch, J F Lange, Y M Bastiaansen-Jenniskens
ABSTRACT
Macrophages play an important role in the reaction to biomaterials, which sometimes have to be used in a surgical field at risk of contamination. The macrophage phenotype in reaction to biomaterials in an inflammatory environment was evaluated in both an in vivo and in vitro setting. In the in vivo setting, polypropylene (PP) biomaterial was implanted for 28 days in the contaminated abdominal wall of rats, and upon removal analysed by routine histology as well as immunohistochemistry for CD68 (marker for macrophages), inducible nitric oxide synthase (iNOS - a marker for proinflammatory M1 macrophages) and CD206 (marker for anti-inflammatory M2 macrophages). For the in vitro model, human peripheral blood monocytes were cultured for 3 days on biomaterials made from PP, collagen (COL), polyethylene terephthalate (PET) and PET coated with collagen (PET+COL). These experiments were performed both with and without lipopolysaccharide and interferon γ stimulation. Secretion of both M1- and M2-related proteins was measured, and a relative M1/M2 index was calculated. In vivo, iNOS- and CD206-positive cells were found around the fibres of the implanted PP biomaterial. In vitro, macrophages on both PP and COL biomaterial had a relatively low M1/M2 index. Macrophages on the PET biomaterial had a high M1/M2 index, with the highest increase of M1 cytokines in an inflammatory environment. Macrophages on the PET+COL biomaterial also had a high M1/M2 index. Macrophages in an inflammatory environment in vitro still react in a biomaterial-dependent manner. This model can help to select biomaterials that are tolerated best in a surgical environment at risk of contamination.
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered