Merck
  • A new stationary phase for high performance liquid chromatography: Calix[4]arene derivatized chitosan bonded silica gel.
A new stationary phase for high performance liquid chromatography: Calix[4]arene derivatized chitosan bonded silica gel.
Journal of chromatography. A (2014-05-28)
Jing Lu, Wenfen Zhang, Yanhao Zhang, Wenjie Zhao, Kai Hu, Ajuan Yu, Pu Liu, Yangjie Wu, Shusheng Zhang
ABSTRACT
In the present paper, a new calix[4]arene derivatized chitosan bonded stationary phase (CCS4) for high performance liquid chromatography (HPLC) was synthesized and characterized. Its chromatographic performance and retention mechanism were evaluated in reversed-phase mode compared with ODS using different solute probes including mono-substituted benzenes, phenols and nucleosides. The results showed that CCS4 stationary phase could provide various interactions with solutes, such as hydrophobic, hydrophilic, π-π, and inclusion interactions. It could perform the mixed-mode separation including RP and HILIC.
Sigma-Aldrich
Chitosan, from shrimp shells, ≥75% (deacetylated)
Sigma-Aldrich
Chitosan, from shrimp shells, practical grade
Sigma-Aldrich
Chitosan, low molecular weight
Sigma-Aldrich
Chitosan, medium molecular weight
Sigma-Aldrich
Chitosan, high molecular weight
Sigma-Aldrich
Chitosan from shrimp shells, low-viscous