Merck
  • Natural dissolved organic matter mobilizes Cd but does not affect the Cd uptake by the green algae Pseudokirchneriella subcapitata (Korschikov) in resin buffered solutions.
Natural dissolved organic matter mobilizes Cd but does not affect the Cd uptake by the green algae Pseudokirchneriella subcapitata (Korschikov) in resin buffered solutions.
Aquatic toxicology (Amsterdam, Netherlands) (2014-05-31)
Liesbeth Verheyen, Liske Versieren, Erik Smolders
ABSTRACT
Natural dissolved organic matter (DOM) can have contrasting effects on metal bioaccumulation in algae because of complexation reactions that reduce free metal ion concentrations and because of DOM adsorption to algal surfaces which promote metal adsorption. This study was set up to reveal the role of different natural DOM samples on cadmium (Cd) uptake by the green algae Pseudokirchneriella subcapitata (Korschikov). Six different DOM samples were collected from natural freshwater systems and isolated by reverse osmosis. In addition, one (13)C enriched DOM sample was isolated from soil to trace DOM adsorption to algae. Algae were exposed to standardized solutions with or without these DOM samples, each exposed at equal DOM concentrations and at equal non-toxic Cd(2+) activity (∼4 nM) that was buffered with a resin. The DOM increased total dissolved Cd by factors 3-16 due to complexation reactions at equal Cd(2+) activity. In contrast, the Cd uptake was unaffected by DOM or increased maximally 1.6 fold ((13)C enriched DOM). The (13)C analysis revealed that maximally 6% of algal C was derived from DOM and that this can explain the small increase in biomass Cd. It is concluded that free Cd(2+) and not DOM-complexed Cd is the main bioavailable form of Cd when solution Cd(2+) is well buffered.
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Cadmium, foil, not light tested, 25x25mm, thickness 0.01mm, 99.7+%
Cadmium, foil, not light tested, 50x50mm, thickness 0.005mm, 99.7+%
Cadmium, wire reel, 0.5m, diameter 0.25mm, hard, 99.99+%
Cadmium, foil, 10mm disks, thickness 0.25mm, as rolled, 99.95%
Cadmium, wire reel, 0.1m, diameter 1.0mm, hard, 99.999%
Cadmium, foil, 0.5m coil, thickness 0.5mm, as rolled, 100%
Cadmium, foil, 25mm disks, thickness 0.125mm, as rolled, 99.95%
Cadmium, foil, 100x100mm, thickness 0.125mm, as rolled, 99.95%
Cadmium, foil, 25mm disks, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, 25mm disks, thickness 0.125mm, as rolled, 99.99+%
Cadmium, foil, 25x25mm, thickness 0.5mm, as rolled, 99.999%
Cadmium, foil, 15mm disks, thickness 0.125mm, as rolled, 99.95%
Cadmium, foil, 100x100mm, thickness 0.125mm, as rolled, 99.99+%
Cadmium, wire reel, 0.2m, diameter 0.5mm, hard, 99.95+%
Cadmium, foil, 15mm disks, thickness 0.25mm, as rolled, 99.9%
Cadmium, foil, 10mm disks, thickness 0.015mm, 99.7+%
Cadmium, foil, 300x300mm, thickness 2.0mm, as rolled, 99.99%
Cadmium, foil, 50x200mm, thickness 1.0mm, as rolled, 99.999%
Cadmium, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99%
Cadmium, wire reel, 0.5m, diameter 0.70mm, as drawn, 99.99+%
Cadmium, foil, 10mm disks, thickness 1.0mm, as rolled, 100%
Cadmium, foil, 50x50mm, thickness 1.0mm, as rolled, 99.95%
Cadmium, foil, 200x200mm, thickness 1.0mm, as rolled, 99.99%
Cadmium, foil, 150x150mm, thickness 0.25mm, as rolled, 99.95%
Cadmium, wire reel, 0.2m, diameter 1.0mm, hard, 99.999%
Cadmium, foil, 10mm disks, thickness 0.25mm, as rolled, 99.9%