Merck
  • Lavender essential oil inhalation suppresses allergic airway inflammation and mucous cell hyperplasia in a murine model of asthma.
Lavender essential oil inhalation suppresses allergic airway inflammation and mucous cell hyperplasia in a murine model of asthma.
Life sciences (2014-06-10)
Tomoe Ueno-Iio, Misako Shibakura, Kanayo Yokota, Michinori Aoe, Tomoko Hyoda, Ryoko Shinohata, Arihiko Kanehiro, Mitsune Tanimoto, Mikio Kataoka
ABSTRACT
Lavender essential oil (Lvn) has been reported to have anti-inflammatory effects. Bronchial asthma is characterized by bronchial allergic inflammation with airway remodeling. Therefore, we evaluated the anti-inflammatory effect of Lvn on experimentally induced bronchial asthma in a murine model. BALB/c mice were sensitized by an intraperitoneal injection of ovalbumin (OVA) at days 0 and 14, and subsequently challenged with nebulized OVA on days 28-30 (Control-Asthma group). Mice in the treatment group inhaled Lvn on days 14-31 (Lvn-Asthma group). The allergic inflammatory response was determined on days 32 and 33. An increase in airway resistance was inhibited in the Lvn-Asthma group than in the Control-Asthma group. The Lvn-Asthma group showed lower total cell numbers and eosinophils in bronchoalveolar lavage (BAL) fluids and peribronchial and perivascular tissues when compared with the Control-Asthma group. The Lvn-Asthma group also had less mucin hyperplasia than the Control-Asthma group. Furthermore, the Lvn-Asthma group showed lower interleukin (IL)-5 and IL-13 cytokine levels in BAL fluids, as well as reduced IL-4 and IL-5 mRNA expression in lung tissue, compared with the Control-Asthma group and determined by FlowCytomix and reverse transcriptase-polymerase chain reaction (RT-PCR), respectively. In addition, Lvn inhalation reduced Muc5b mRNA expression in the lungs without significantly changing the expression of Muc5ac mRNA. Lvn inhibits allergic inflammation and mucous cell hyperplasia with suppression of T-helper-2 cell cytokines and Muc5b expression in a murine model of asthma. Consequently, Lvn may be useful as an alternative medicine for bronchial asthma.
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5%
Supelco
Albumin from chicken egg white, For use as a marker in SDS-PAGE
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from chicken egg white, powder, 62-88% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥90% (agarose gel electrophoresis)
Sigma-Aldrich
Lavender oil, natural, from Lavandula angustifolia L.
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioXtra, ≥98.0%
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, ACS reagent, ≥98%
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Lavender oil 40/42% fleurs, natural, FG