Merck
  • Study on dioxygen reduction by mutational modifications of the hydrogen bond network leading from bulk water to the trinuclear copper center in bilirubin oxidase.
Study on dioxygen reduction by mutational modifications of the hydrogen bond network leading from bulk water to the trinuclear copper center in bilirubin oxidase.
Biochemical and biophysical research communications (2014-06-22)
Hirotoshi Morishita, Daisuke Kurita, Kunishige Kataoka, Takeshi Sakurai
ABSTRACT
The hydrogen bond network leading from bulk water to the trinuclear copper center in bilirubin oxidase is constructed with Glu463 and water molecules to transport protons for the four-electron reduction of dioxygen. Substitutions of Glu463 with Gln or Ala were attributed to virtually complete loss or significant reduction in enzymatic activities due to an inhibition of the proton transfer steps to dioxygen. The single turnover reaction of the Glu463Gln mutant afforded the highly magnetically interacted intermediate II (native intermediate) with a broad g=1.96 electron paramagnetic resonance signal detectable at cryogenic temperatures. Reactions of the double mutants, Cys457Ser/Glu463Gln and Cys457Ser/Glu463Ala afforded the intermediate I (peroxide intermediate) because the type I copper center to donate the fourth electron to dioxygen was vacant in addition to the interference of proton transport due to the mutation at Glu463. The intermediate I gave no electron paramagnetic resonance signal, but the type II copper signal became detectable with the decay of the intermediate I. Structural and functional similarities between multicopper oxidases are discussed based on the present mutation at Glu463 in bilirubin oxidase.
Product Description
Copper, wire reel, 50m, diameter 0.15mm, annealed, silver plated, 99.9%
Glutamic acid, European Pharmacopoeia (EP) Reference Standard
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Hydrogen, ≥99.999%
Copper - O.F.H.C., foil, 150x150mm, thickness 0.25mm, annealed, 99.95+%
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Copper, rod, 100mm, diameter 16mm, as drawn, 99.99+%
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
L-Glutamic acid hydrochloride, ≥99% (HPLC)
Copper - O.F.H.C., foil, 25x25mm, thickness 0.25mm, annealed, 99.95+%
Copper, wire reel, 500m, diameter 0.5mm, annealed, 99.9%
Copper - O.F.H.C., foil, 100x100mm, thickness, 0.5mm, half hard, flattened, polished on both sides, 99.95+%
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Copper, wire reel, 1000m, diameter 0.125mm, annealed, 99.9%
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Copper, wire reel, 1m, diameter 0.01mm, hard, 99.99%
Copper - O.F.H.C., foil, light tested, 150x150mm, thickness 0.05mm, annealed, 99.95+%
Copper, rod, 100mm, diameter 4.8mm, hard, 99.9%
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Copper, wire reel, 200m, diameter 0.5mm, annealed, silver plated, 99.9%
Copper - O.F.H.C., wire reel, 10m, diameter 0.25mm, as drawn, 99.95+%
Copper, wire reel, 5m, diameter 0.05mm, as drawn, 99.99+%
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Copper - O.F.H.C., tube, 500mm, outside diameter 4.0mm, inside diameter 3mm, wall thickness 0.5mm, 99.95+%