Merck
  • Preclinical gastrointestinal prokinetic efficacy and endocrine effects of the ghrelin mimetic RM-131.
Preclinical gastrointestinal prokinetic efficacy and endocrine effects of the ghrelin mimetic RM-131.
Life sciences (2014-06-17)
Lex Van der Ploeg, Haley Laken, Shubh Sharma, Rakesh Datta, Heather Halem, Jesse Dong, Caroline Touvay, Marc Teillot, Patrick Noonan, Lou Tartaglia, Liz Stoner, Bart Henderson, Keith Gottesdiener, Michael Culler
ABSTRACT
The 28 amino acid hormone ghrelin, the natural ligand for the growth hormone secretagogue, or ghrelin receptor (GHR), has diverse physiological functions, including a possible role as a gastrointestinal prokinetic. The synthetic ghrelin mimetic RM-131 is in Phase II clinical trials for treatment of diabetic gastroparesis and other gastrointestinal (GI) disorders. We aimed to determine the relative potency of RM-131, when compared to other GI ghrelin mimetics, to predict efficacy and determine the role of RM-131 in models of inflammatory bowel disease. We evaluated and compared ghrelin, RM-131 and other synthetic ghrelin mimetics for their prokinetic potency in models of gastrointestinal disorders in the rat and we evaluated the endocrine (rats and dogs) and anti-inflammatory effects (mice) of the ghrelin mimetic RM-131. The pentapeptide RM-131 increased gastric emptying in rodent models of ileus. RM-131 is about 100-fold more potent than human ghrelin and is 600 to 1800-fold more potent, when compared to several investigational ghrelin mimetics tested in clinical trials. RM-131 has anti-inflammatory effects and significantly increases survival and reduces macroscopic markers of tissue damage in a TNBS model of inflammatory bowel disease. RM-131 treatment shows a transient increase in growth hormone levels in Beagle dogs and rats, returning to baseline upon chronic treatment. Significant effects on glucose and insulin are not observed in chronic studies. RM-131's potency, efficacy and endocrine profile, are promising attributes for the treatment of diverse functional gastrointestinal disorders in humans.
Product Description
USP
Phenol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phenol, natural, 97%, FG
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Supelco
Phenol, PESTANAL®, analytical standard