Merck
  • Comparison of two ionic liquid dispersive liquid-liquid microextraction approaches for the determination of benzoylurea insecticides in wastewater using liquid chromatography-quadrupole-linear ion trap-mass spectrometry: evaluation of green parameters.
Comparison of two ionic liquid dispersive liquid-liquid microextraction approaches for the determination of benzoylurea insecticides in wastewater using liquid chromatography-quadrupole-linear ion trap-mass spectrometry: evaluation of green parameters.
Journal of chromatography. A (2014-07-06)
M M Parrilla Vázquez, P Parrilla Vázquez, M Martínez Galera, A Uclés Moreno
ABSTRACT
Two dispersive liquid-liquid microextraction (DLLME) approaches including temperature-controlled ionic liquid dispersive liquid-liquid microextraction (TCIL-DLLME) and ultrasound-assisted ionic liquid dispersive liquid-liquid microextraction (US-IL-DLLME) were compared for the extraction of six benzoylurea insecticides (diflubenzuron, triflumuron, hexaflumuron, teflubenzuron, lufenuron and flufenoxuron) from wastewater samples prior to their determination by high-performance liquid chromatography with a hybrid triple quadrupole-linear ion trap-mass spectrometer (LC-QqLIT-MS/MS). Influential parameters affecting extraction efficiency were systematically studied and optimized and the most significant green parameters were quantified and compared. The best results were obtained using the US-IL-DLLME procedure, which employed the IL 1-octyl-3-methylimidazolium hexafluorophosphate ([C8MIM][PF6]) and methanol (MeOH) as extraction and disperser solvent, respectively. US-IL-DLLME procedure was fast, easy, low environmental toxicity and, it was also able to successfully extract all selected benzoylureas. This method was extensively validated with satisfactory results: limits of detection and quantification were in the range 0.5-1.0 ng L(-1) and 1.5-3.5 ng L(-1), respectively, whereas recovery rates ranged from 89 to 103% and the relative standard deviations were lower than 13.4%. The applicability of the method was assessed with the analysis of effluent wastewater samples from a wastewater treatment plant located in an agricultural zone of Almería (Spain) and the results indicated the presence of teflubenzuron at mean concentration levels of 11.3 ng L(-1). US-IL-DLLME sample treatment in combination with LC-QqLIT-MS/MS has demonstrated to be a sensitive, selective and efficient method to determine benzoylurea insecticides in wastewaters at ultra-trace levels.
Product Description
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Hexaflumuron, PESTANAL®, analytical standard
Supelco
Flufenoxuron, PESTANAL®, analytical standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Supelco
Teflubenzuron, PESTANAL®, analytical standard
Sigma-Aldrich
1-Butyl-3-methylimidazolium hexafluorophosphate, for catalysis, ≥98.5% (T)
Supelco
Lufenuron, PESTANAL®, analytical standard
Lufenuron, European Pharmacopoeia (EP) Reference Standard
Supelco
Triflumuron, PESTANAL®, analytical standard
Supelco
Diflubenzuron, PESTANAL®, analytical standard
Sigma-Aldrich
1-Butyl-3-methylimidazolium hexafluorophosphate, ≥97.0% (HPLC)
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, analytical standard