Merck
  • Estimates of projection overlap and zones of convergence within frontal-striatal circuits.
Estimates of projection overlap and zones of convergence within frontal-striatal circuits.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2014-07-18)
Bruno B Averbeck, Julia Lehman, Moriah Jacobson, Suzanne N Haber
ABSTRACT
Frontal-striatal circuits underlie important decision processes, and pathology in these circuits is implicated in many psychiatric disorders. Studies have shown a topographic organization of cortical projections into the striatum. However, work has also shown that there is considerable overlap in the striatal projection zones of nearby cortical regions. To characterize this in detail, we quantified the complete striatal projection zones from 34 cortical injection locations in rhesus monkeys. We first fit a statistical model that showed that the projection zone of a cortical injection site could be predicted with considerable accuracy using a cross-validated model estimated on only the other injection sites. We then examined the fraction of overlap in striatal projection zones as a function of distance between cortical injection sites, and found that there was a highly regular relationship. Specifically, nearby cortical locations had as much as 80% overlap, and the amount of overlap decayed exponentially as a function of distance between the cortical injection sites. Finally, we found that some portions of the striatum received inputs from all the prefrontal regions, making these striatal zones candidates as information-processing hubs. Thus, the striatum is a site of convergence that allows integration of information spread across diverse prefrontal cortical areas.
Product Description
Dextran 70 for calibration, Dextran 70 European Pharmacopoeia (EP) Reference Standard
Dextran 10 for calibration, Dextran 10 European Pharmacopoeia (EP) Reference Standard
Dextran 40 for calibration, Dextran 40 European Pharmacopoeia (EP) Reference Standard
Dextran 250 for calibration, Dextran 250 European Pharmacopoeia (EP) Reference Standard
Dextran 40 for performance test, Dextran 40 European Pharmacopoeia (EP) Reference Standard
Dextran 60/70 for performance test, Dextran 60/70 European Pharmacopoeia (EP) Reference Standard
Dextran EP Reference Standard, European Pharmacopoeia (EP) Reference Standard
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, suitable for gel permeation chromatography (GPC), Mw 25,000
Supelco
Dextran, analytical standard, suitable for gel permeation chromatography (GPC), Set Mp 1,000-400,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 9,000-11,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 15,000-30,000
Dextran 4 for calibration, Dextran 4 European Pharmacopoeia (EP) Reference Standard
Dextran 1, Dextran 1 European Pharmacopoeia (EP) Reference Standard
Dextran Vo, Dextran Vo European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 60,000-76,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, suitable for gel permeation chromatography (GPC), Mw 50,000
Supelco
Dextran, analytical standard, 25,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 150,000
Supelco
Dextran, analytical standard, 12,000
Supelco
Dextran, analytical standard, 410,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 450,000-650,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, suitable for gel permeation chromatography (GPC), Mw 5,000
Supelco
Dextran, analytical standard, 1,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~6,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~200,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~70,000
Sigma-Aldrich
Dextran solution from Leuconostoc mesenteroides, 20 % (w/w) (Autoclaved)
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~40,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 35,000-45,000