Merck
  • Molecular characterization of a Chinese family carrying a novel C4329A mutation in mitochondrial tRNAIle and tRNAGln genes.
Molecular characterization of a Chinese family carrying a novel C4329A mutation in mitochondrial tRNAIle and tRNAGln genes.
BMC medical genetics (2014-07-25)
Yuqi Liu, Yang Li, Jinliao Gao, Chao Zhu, Yunfeng Lan, Jie Yang, Zongbin Li, Minxin Guan, Yundai Chen
ABSTRACT
Hypertension is a very common cardiovascular disease influenced by multiple genetic and environmental factors. More recently, there are some studies showed that mutations in mitochondrial DNA have been involved in its pathogenesis. In this study we did further investigations on this relationship. Epidemiological research found a Han Chinese family with probable maternally transmitted hypertension. Sequence analysis of the whole mitochondrial DNA was detected from all the family members. And evaluations of the clinical, genetic and molecular characterization were also performed. Matrilineal relatives within the family exhibited varying degrees of hypertension with an onset age of 48-55 years. Sequence analysis of this pedigree showed a novel homoplasmic 4329C > G mutation located at the 3' end of the tRNAIle and tRNAGln genes that was absent from 366 Chinese controls. The cytosine (C) at 4329 position was very important in the structural formation and stabilization of functional tRNAs, which was highly conserved in mitochondria of various organisms and also contributed to the high fidelity of the acceptor arm. Cells carrying this mutation were also shown to harbor mitochondrial dysfunctions. The C4329G point mutation in tRNAIle and tRNAGln was involved in the pathogenesis of hypertension, perhaps in association with other modifying factors.
Product Description
Supelco
Cytosine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Guanine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Guanine, 98%
Sigma-Aldrich
Guanine, BioUltra
Sigma-Aldrich
Cytosine, ≥99%
Gemcitabine impurity A, European Pharmacopoeia (EP) Reference Standard