Merck
  • Short-term effect of ascorbate on bacterial content, plasminogen activator inhibitor-1, and myeloperoxidase in septic mice.
Short-term effect of ascorbate on bacterial content, plasminogen activator inhibitor-1, and myeloperoxidase in septic mice.
The Journal of surgical research (2014-05-13)
Scott B Swarbreck, Dan Secor, Christopher G Ellis, Michael D Sharpe, John X Wilson, Karel Tyml
ABSTRACT
Sepsis, a potential risk associated with surgery, leads to a systemic inflammatory response including the plugging of capillary beds. This plugging may precipitate organ failure and subsequent death. We have shown that capillary plugging can be reversed rapidly within 1 h by intravenous injection of ascorbate in mouse skeletal muscle. It is unknown whether, in parallel with this effect, ascorbate negatively affects the protective responses to sepsis involving the fibrinolytic and immune systems. We hypothesized that treatment with ascorbate for 1 h does not alter bacterial content, plasminogen activator inhibitor 1 (PAI-1), and neutrophil infiltration in lung, kidney, spleen, and liver (organs with high immune response) of septic mice. Sepsis was induced by feces injection into the peritoneum. Mice were injected intravenously with ascorbate at 6 h (10 mg/kg), and samples of peritoneal fluid, arterial blood, and organs collected at 7 h were subjected to analyses of bacterial content, PAI-1 messenger RNA and enzymatic activity, and myeloperoxidase (MPO) (a measure of neutrophil infiltration). Sepsis increased bacterial content in all fluids and organs and increased PAI-1 messenger RNA and enzymatic activity in the lung and liver. Sepsis increased the myeloperoxidase level in the lung and liver, and lowered it in the spleen. Except for decreasing the bacterial content in blood, these responses to sepsis were not altered by ascorbate. The rapid effect of ascorbate against capillary plugging in the septic mouse skeletal muscle is not accompanied by alterations in PAI-1 or myeloperoxidase responses in the organs with high immune response.
Product Description
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sodium ascorbate, European Pharmacopoeia (EP) Reference Standard
Supelco
Sodium ascorbate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%