Merck
  • Chronic sensitivity of white sturgeon (Acipenser transmontanus) and rainbow trout (Oncorhynchus mykiss) to cadmium, copper, lead, or zinc in laboratory water-only exposures.
Chronic sensitivity of white sturgeon (Acipenser transmontanus) and rainbow trout (Oncorhynchus mykiss) to cadmium, copper, lead, or zinc in laboratory water-only exposures.
Environmental toxicology and chemistry (2014-05-28)
Ning Wang, Christopher G Ingersoll, Rebecca A Dorman, William G Brumbaugh, Christopher A Mebane, James L Kunz, Doug K Hardesty
ABSTRACT
Chronic toxicity of cadmium, copper, lead, or zinc to white sturgeon (Acipenser transmontanus) and rainbow trout (Oncorhynchus mykiss) was evaluated in water-only exposures started with newly hatched larvae or approximately 1-mo-old juveniles. The 20% effect concentration (EC20) for cadmium from the sturgeon tests was higher than the EC20 from the trout tests, whereas the EC20 for copper, lead, or zinc for the sturgeon were lower than those EC20s for the trout. When the EC20s from the present study were included in compiled toxicity databases for all freshwater species, species mean chronic value for white sturgeon was in a relatively low percentile of the species sensitivity distribution for copper (9th percentile) and in the middle percentile for cadmium (55th percentile), zinc (40th percentile), or lead (50th percentile). However, the species mean chronic value for rainbow trout was in a high percentile for copper, lead, and zinc (∼68th-82nd percentile), but in a low percentile for cadmium (23rd percentile). The trout EC20s for each of the 4 metals and the sturgeon EC20s for cadmium or lead were above US Environmental Protection Agency chronic ambient water quality criteria (AWQC) or Washington State chronic water quality standards (WQS), whereas the sturgeon EC20s for copper or zinc were approximately equal to or below the chronic AWQC and WQS. In addition, acute 50% effect concentrations (EC50s) for copper obtained in the first 4 d of the chronic sturgeon test were below the final acute value used to derive acute AWQC and below acute WQS for copper.
Product Description
Copper, wire reel, 50m, diameter 0.15mm, annealed, silver plated, 99.9%
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn2+ in THF, highly reactive Rieke®metal
Continuous cast copper (O), BCR®, certified reference material, rod
Copper - O.F.H.C., foil, 150x150mm, thickness 0.25mm, annealed, 99.95+%
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Copper, rod, 100mm, diameter 16mm, as drawn, 99.99+%
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Copper - O.F.H.C., foil, 25x25mm, thickness 0.25mm, annealed, 99.95+%
Copper, wire reel, 500m, diameter 0.5mm, annealed, 99.9%
Copper - O.F.H.C., foil, 100x100mm, thickness, 0.5mm, half hard, flattened, polished on both sides, 99.95+%
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Copper, wire reel, 1000m, diameter 0.125mm, annealed, 99.9%
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Copper, wire reel, 1m, diameter 0.01mm, hard, 99.99%
Copper - O.F.H.C., foil, light tested, 150x150mm, thickness 0.05mm, annealed, 99.95+%
Copper, rod, 100mm, diameter 4.8mm, hard, 99.9%
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Copper, wire reel, 200m, diameter 0.5mm, annealed, silver plated, 99.9%
Copper - O.F.H.C., wire reel, 10m, diameter 0.25mm, as drawn, 99.95+%
Copper, wire reel, 5m, diameter 0.05mm, as drawn, 99.99+%
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Zinc, foil, 1m coil, thickness 0.075mm, as rolled, 99.95+%
Zinc, wire reel, 1m, diameter 1.0mm, 99.99+%
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Zinc, wire reel, 100m, diameter 1.0mm, extruded, 99.9%
Zinc, foil, 1m coil, thickness 0.125mm, as rolled, 99.95+%
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%