Merck
  • Histopathological features and their prognostic impact in nodular lymphocyte-predominant Hodgkin lymphoma--a matched pair analysis from the German Hodgkin Study Group (GHSG).
Histopathological features and their prognostic impact in nodular lymphocyte-predominant Hodgkin lymphoma--a matched pair analysis from the German Hodgkin Study Group (GHSG).
British journal of haematology (2014-06-27)
Sylvia Hartmann, Dennis A Eichenauer, Annette Plütschow, Anja Mottok, Roshanak Bob, Karoline Koch, Heinz-Wolfram Bernd, Sergio Cogliatti, Michael Hummel, Alfred C Feller, German Ott, Peter Möller, Andreas Rosenwald, Harald Stein, Martin-Leo Hansmann, Andreas Engert, Wolfram Klapper
ABSTRACT
Nodular lymphocyte-predominant Hodgkin lymphoma (NLPHL) is a rare lymphoma entity. We performed a matched-pair analysis to evaluate the prognostic impact of several histopathological features in this distinct Hodgkin lymphoma subtype. Lymph node samples of NLPHL patients were tested for CD15, IgD, phosphorylated STAT6, ICOS and Epstein-Barr virus status of the malignant lymphocyte-predominant cells as well as epithelioid cell clusters and activated T cells in the microenvironment. None of these features was associated with a particular clinical outcome. However, patients presenting with epithelioid cell clusters showed a non-significant trend towards a lower relapse rate, justifying further evaluation of this marker.
Sigma-Aldrich
Phosphorylase b from rabbit muscle, lyophilized powder, ≥20 units/mg protein, 2× crystallization
Sigma-Aldrich
Phosphorylase b from rabbit muscle, For use as a marker in SDS-PAGE