Merck
  • Electrochemical degradation of nitrobenzene by anodic oxidation on the constructed TiO2-NTs/SnO2-Sb/PbO2 electrode.
Electrochemical degradation of nitrobenzene by anodic oxidation on the constructed TiO2-NTs/SnO2-Sb/PbO2 electrode.
Chemosphere (2014-07-30)
Yong Chen, Hongyi Li, Weijing Liu, Yong Tu, Yaohui Zhang, Weiqing Han, Lianjun Wang
ABSTRACT
The interlayer of Sb-doped SnO2 (SnO2-Sb) and TiO2 nanotubes (TiO2-NTs) on Ti has been introduced into the PbO2 electrode system with the aim to reveal the mechanism of enhanced electrochemical performance of TiO2-NTs/SnO2-Sb/PbO2 electrode. In contrast with the traditional Ti/SnO2-Sb/PbO2 electrode, the constructed PbO2 electrode has a more regular and compact morphology with better oriented crystals of lower size. The TiO2-NTs/SnO2-Sb interlayer prepared by electrodeposition process improves PbO2 coating structure effectively, and enhances the electrochemical performance of PbO2 electrode. Kinetic analyses indicated that the electrochemical oxidation of nitrobenzene on the PbO2 electrodes followed pseudo-first-order reaction, and mass transport was enhanced at the constructed electrode. The accumulation of nitrocompounds of degradation intermediates on constructed electrode was lower, and almost all of the nitro groups were eliminated from aromatic rings after 6h of electrolysis. Higher combustion efficiency was obtained on the constructed TiO2-NTs/SnO2-Sb/PbO2 electrode. The intermediates of nitrobenzene oxidation were confirmed by IC and GC/MS.
Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Titanium, ≤10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Sigma-Aldrich
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Titanium, IRMM®, certified reference material, 0.5 mm wire
Titanium, IRMM®, certified reference material, 0.5 mm foil
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, 99.8% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Titanium, wire reel, 10m, diameter 0.125mm, annealed, 99.6+%
Lead, foil, 100x100mm, thickness 1.5mm, as rolled, 99.99+%
Titanium, wire reel, 1m, diameter 0.5mm, as drawn, 99.99+%
Lead, foil, 100x100mm, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 5m coil, thickness 0.1mm, as rolled, 99.95%
Titanium, wire reel, 25m, diameter 0.25mm, as drawn, 99.6+%
Lead, foil, 6mm disks, thickness 0.25mm, as rolled, 99.99+%
Titanium, tube, 500mm, outside diameter 2.03mm, inside diameter 1.63mm, wall thickness 0.2mm, annealed, 99.6+%
Lead, foil, 10mm disks, thickness 0.008mm, 99.99+%
Lead, foil, 8mm disks, thickness 0.015mm, 99.99+%
Lead, foil, 4mm disks, thickness 0.04mm, 99.99+%
Lead, foil, 500x500mm, thickness 3.0mm, as rolled, 99.95%
Lead, foil, 25x25mm, thickness 0.55mm, as rolled, 99.999%
Lead, foil, 150x150mm, thickness 0.55mm, as rolled, 99.999%
Lead, foil, 6mm disks, thickness 0.55mm, as rolled, 100%
Titanium, wire reel, 0.5m, diameter 0.25mm, as drawn, 99.99+%
Lead, foil, light tested, 100x100mm, thickness 0.025mm, 99.99+%