Merck
  • Effect of anions and humic acid on the performance of nanoscale zero-valent iron particles coated with polyacrylic acid.
Effect of anions and humic acid on the performance of nanoscale zero-valent iron particles coated with polyacrylic acid.
Chemosphere (2014-07-30)
Hong-Seok Kim, Jun-Young Ahn, Cheolyong Kim, Seockheon Lee, Inseong Hwang
ABSTRACT
Effects of anions (NO3(-), HCO3(-), Cl(-), SO4(2-)) and humic acid on the reactivity and core/shell chemistries of polyacrylic acid-coated nanoscale zero-valent iron (PAA-NZVI) and inorganically modified NZVI (INORG-NZVI) particles were investigated. The reactivity tests under various ion concentrations (0.2-30mN) revealed the existence of a favorable molar ratio of anion/NZVI that increased the reactivity of NZVI particles. The presence of a relatively small amount of humic acid (0.5mgL(-1)) substantially decreased the INORG-NZVI reactivity by 76%, whereas the reactivity of PAA-NZVI decreased only by 12%. The XRD and TEM results supported the role of the PAA coating of PAA-NZVI in impeding the oxidation of the Fe(0) core by groundwater solutes. This protective role provided by the organic coating also resulted in a 2.3-fold increase in the trichloroethylene (TCE) reduction capacity of PAA-NZVI compared to that of INORG-NZVI in the presence of anions/humic acid. Ethylene and ethane were simultaneously produced as the major reduction products of TCE in both NZVI systems, suggesting that a hydrodechlorination occurred without the aid of metallic catalysts. The PAA coating, originally designed to improve the mobility of NZVI, enhanced TCE degradation performances of NZVI in the presence of anions and humic acid.
Sigma-Aldrich
Carbonyl iron, ≥97% Fe basis
Iron, rod, 1000mm, diameter 6.35mm, as drawn, soft ingot 99.8%
USP
Residual Solvent Class 2 - Trichloroethylene, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Iron, foil, thickness 0.25 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Iron, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, granular, 10-40 mesh, >99.99% trace metals basis
Iron, rod, 200mm, diameter 80mm, as drawn, armcO« soft ingot 99.8%
Iron, foil, not light tested, 50x50mm, thickness 0.004mm, 99.85%
Iron, foil, 25x25mm, thickness 1.0mm, as rolled, 99.99+%
Iron, foil, 2m coil, thickness 0.5mm, hard, 99.5%
Sigma-Aldrich
Iron, ≥99%, reduced, powder (fine)
Iron, foil, not light tested, 25x25mm, thickness 0.005mm, 99.85%
Iron, foil, 150x150mm, thickness 3.0mm, as rolled, 99.5%
Iron, foil, 0.5m coil, thickness 0.05mm, hard, 99.5%
Iron, foil, 10m coil, thickness 0.15mm, hard, 99.5%
Iron, foil, 50x50mm, thickness 1.83mm, Quarter hard, 99.5%
Iron, tube, 100mm, outside diameter 5.0mm, inside diameter 4.5mm, wall thickness 0.25mm, as drawn, 99.5%
Iron, wire reel, 2m, diameter 0.13mm, hard, 99.99+%
Iron, wire reel, 10m, diameter 0.065mm, hard, 99.5%
Iron, tube, 500mm, outside diameter 4.0mm, inside diameter 3.6mm, wall thickness 0.2mm, as drawn, 99.5%
Iron, wire reel, 5m, diameter 0.13mm, hard, 99.99+%
Iron, tube, 100mm, outside diameter 2.0mm, inside diameter 1.08mm, wall thickness 0.46mm, as drawn, 99.5%
Iron, foil, 25x25mm, thickness 2.0mm, as rolled, 99.95%
Iron, wire reel, 0.5m, diameter 0.5mm, hard, 99.99+%
Iron, tube, 1000mm, outside diameter 4.0mm, inside diameter 3.6mm, wall thickness 0.2mm, as drawn, 99.5%
Iron, tube, 500mm, outside diameter 20.0mm, inside diameter 16mm, wall thickness 2.0mm, annealed, 99.5%
Iron, foil, 10mm disks, thickness 0.004mm, 99.85%
Iron, tube, 200mm, outside diameter 3.2mm, inside diameter 2.2mm, wall thickness 0.5mm, as drawn, 99.8%
Iron, foil, 0.2m coil, thickness 0.5mm, coil width 49mm, armco« soft ingot 99.8+%
Iron, wire reel, 10m, diameter 0.5mm, as drawn, 99.95%