Merck
  • Dietary fiber supplementation for fecal incontinence: a randomized clinical trial.
Dietary fiber supplementation for fecal incontinence: a randomized clinical trial.
Research in nursing & health (2014-08-27)
Donna Z Bliss, Kay Savik, Hans-Joachim G Jung, Robin Whitebird, Ann Lowry, Xiaoyan Sheng
ABSTRACT
Dietary fiber supplements are used to manage fecal incontinence (FI), but little is known about the fiber type to recommend or the level of effectiveness of such supplements, which appears related to the fermentability of the fiber. The aim of this single-blind, randomized controlled trial was to compare the effects of three dietary fiber supplements (carboxymethylcellulose [CMC], gum arabic [GA], or psyllium) with differing levels of fermentability to a placebo in community-living individuals incontinent of loose/liquid feces. The primary outcome was FI frequency; secondary outcomes included FI amount and consistency, supplement intolerance, and quality of life (QoL). Possible mechanisms underlying supplement effects were also examined. After a 14-day baseline, 189 subjects consumed a placebo or 16 g total fiber/day of one of the fiber supplements for 32 days. FI frequency significantly decreased after psyllium supplementation versus placebo, in both intent-to-treat and per-protocol mixed model analyses. CMC increased FI frequency. In intent-to-treat analysis, the number of FI episodes/week after supplementation was estimated to be 5.5 for Placebo, 2.5 for Psyllium, 4.3 for GA, and 6.2 for CMC. Only psyllium consumption resulted in a gel in feces. Supplement intolerance was low. QoL scores did not differ among groups. Patients with FI may experience a reduction in FI frequency after psyllium supplementation, and decreased FI frequency has been shown to be an important personal goal of treatment for patients with FI. Formation of a gel in feces appears to be a mechanism by which residual psyllium improved FI.
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 1.2
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.7
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.9
Sigma-Aldrich
Sodium carboxymethyl cellulose, viscosity 50-200 cP , c=4% H2O at 25­°C
Sigma-Aldrich
Sodium carboxymethyl cellulose, viscosity 2500-6000 cP 
Sigma-Aldrich
Carboxymethylcellulose sodium, meets USP testing specifications, Medium viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, low viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, ultra high viscosity, highly purified
Sigma-Aldrich
Carboxymethylcellulose sodium salt, Medium viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, medium viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, High viscosity