Merck
  • Separation and characterization of underivatized oligosaccharides using liquid chromatography and liquid chromatography-electrospray ionization mass spectrometry.
Separation and characterization of underivatized oligosaccharides using liquid chromatography and liquid chromatography-electrospray ionization mass spectrometry.
Journal of chromatography. A (2005-07-26)
Ying Liu, Sameer Urgaonkar, John G Verkade, Daniel W Armstrong
ABSTRACT
Native cyclodextrin-based columns are particularly useful for the analysis of oligosaccharides because the retention of these carbohydrates is based mainly on the hydrogen bonding interactions of oligosaccharide hydroxyl groups with the stationary phase. Thus, the retention time predictably increases with the number of analyte hydroxyl groups, which corresponds to the elongation of the oligosaccharide chain. High-performance liquid chromatography (HPLC) coupled to electrospray ionization (ESI) mass spectrometry (MS) was used for the separation and characterization of underivatized oligosaccharide mixtures. With the limits of detection as low as 50 pg, all individual components of oligosaccharide mixtures (up to 11 glucose units long) were baseline resolved on a Cyclobond I 2000 column and detected using ESI-MS. Low flow rates and narrow I.D. columns increase the ESI-MS sensitivity significantly. The method showed potential usefulness for the sensitive and quick analysis of hydrolysis products of polysaccharides, and for trace levels of individual oligosaccharide or oligosaccharide isomers from biological systems.
Product Description
Supelco
Astec® CYCLOBOND I 2000 Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Astec® CYCLOBOND I 2000 Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Astec® CYCLOBOND I 2000 Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Astec® CYCLOBOND I 2000 Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm