Merck
  • Efficacy of agents counteracting hypoxia in fractionated radiation regimes.
Efficacy of agents counteracting hypoxia in fractionated radiation regimes.
Radiotherapy and oncology : journal of the European Society for Therapeutic Radiology and Oncology (1996-11-01)
S Stern, M Guichard
ABSTRACT
Solid tumours contain hypoxic cells which are resistant to radiotherapy. This study compares the efficacy of several strategies to counteract diffusion-limited hypoxia, or intermittent hypoxia in a fractionated regimen of 1 to 6 x 2 Gy. Nicotinamide (250 mg/kg), perflubron emulsion (Oxygent) (4 ml/kg), tirapazamine (SR4233) (0.10 mmol/kg) and carbogen breathing, administered alone or in combination, were investigated on two tumour cell lines: EMT6 (a rodent mammary carcinoma) and HRT18 (a human rectal adenocarcinoma) using a clonogenic assay. The radiosensitizing effect of the agents was assessed after 1 and 6 x 2 Gy for drugs used alone, and 1, 2, 4, 6 x 2 Gy for drugs used in combination. At the end of the fractionated radiation regimen, the combination of nicotinamide + carbogen induced the greatest radiosensitization for EMT6 tumours, while greatest radiosensitization of HRT18 was obtained with nicotinamide + carbogen + tirapazamine. The efficacy of the strategies for overcoming hypoxia using a fractionated regimen depends on the tumour cell line. These differences could be linked to differences in the initial percentages of acute and chronic hypoxic cells, and to changes in the two types of hypoxia during treatment.
Sigma-Aldrich
3,9-Divinyl-2,4,8,10-tetraoxaspiro[5.5]undecane, 98%
Pricing and availability is not currently available.