Merck
Biotransformation of 6,6-Dimethylfulvene by Pseudomonas putida RE213.
Applied and environmental microbiology (1996-03-01)
R W Eaton, S A Selifonov
ABSTRACT
The biotransformation of 6,6-dimethylfulvene [5-(1-methylethylidene)-1,3-cyclopentadiene], a nonaromatic C(inf5) carbocyclic analog of isopropylbenzene, was examined by using Pseudomonas putida RE213, a Tn5-generated dihydrodiol-accumulating mutant of the isopropylbenzene-degrading strain P. putida RE204. 6,6-Dimethylfulvene was converted to a single chiral product identified as (+)-(1R,2S)-cis-1,2-dihydroxy-5-(1-methylethylidene)-3-cyclopentene. This isopropylbenzene 2,3-dioxygenase-catalyzed transformation demonstrates the potential of bacterial arene dioxygenases for the direct conversion of cyclopentadienylidene compounds to homochiral C(inf5) carbocyclic cis-diols for use in enantiocontrolled organic syntheses.
Sigma-Aldrich
6,6-Dimethylfulvene, ≥95%
Pricing and availability is not currently available.