Merck
  • Secretion and immunogenicity of the meningioma-associated antigen TXNDC16.
Secretion and immunogenicity of the meningioma-associated antigen TXNDC16.
Journal of immunology (Baltimore, Md. : 1950) (2014-08-15)
Christian Harz, Nicole Ludwig, Sven Lang, Tamara V Werner, Valentina Galata, Christina Backes, Katja Schmitt, Ruth Nickels, Elmar Krause, Martin Jung, Jens Rettig, Andreas Keller, Michael Menger, Richard Zimmermann, Eckart Meese
ABSTRACT
In a previous study, we identified thioredoxin domain containing 16 (TXNDC16) as a meningioma-associated Ag by protein macroarray screening. Serological screening detected autoantibodies against TXNDC16 exclusively in meningioma patients' sera and not in sera of healthy controls. TXNDC16 was previously found to be an endoplasmic reticulum (ER)-luminal glycoprotein. In this study, we show an additional ER-associated localization of TXNDC16 in the cytosol by in vitro synthesis, molecular mass shift assay, and flow cytometry. We were able to show TXNDC16 secretion in different human cell lines due to masked and therefore nonfunctional ER retrieval motif. A previously indicated exosomal TXNDC16 secretion could not be confirmed in HEK293 cells. The secreted serum protein TXNDC16 is bound in circulating immune complexes, which were found both in meningioma and healthy blood donor sera. Employing a customized array with 163 overlapping TXNDC16 peptides and measuring autoantibody reactivity, we achieved discrimination of meningioma sera from healthy controls with an accuracy of 87.2% using a set of only five immunogenic TXNDC16 epitopes.
Sigma-Aldrich
Anti-c-Myc antibody produced in rabbit, ~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-c-Myc antibody, Mouse monoclonal, clone 9E10, purified from hybridoma cell culture
Sigma-Aldrich
Anti-TXNDC16 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-CD63 antibody produced in rabbit, affinity isolated antibody, Prestige Antibodies® Powered by Atlas Antibodies, buffered aqueous glycerol solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)