Merck
  • Severe mandibular atrophy treated with a subperiosteal implant and simultaneous graft with rhBMP-2 and mineralized allograft: a case report.
Severe mandibular atrophy treated with a subperiosteal implant and simultaneous graft with rhBMP-2 and mineralized allograft: a case report.
The Journal of oral implantology (2013-04-12)
Cosimo Loperfido, Juan Mesquida, Jaime L Lozada
ABSTRACT
A 71-year-old patient was successfully rehabilitated by means of a 3D model-derived, hydroxyapatite-coated titanium subperiosteal mandibular implant. The implant was specifically designed to allow bone augmentation. The deficient bone was simultaneously grafted with mineralized bone allograft and recombinant bone morphogenetic protein -2 (rhBMP-2). The 32-month postoperative cone beam computerized tomography follow-up showed vertical bone augmentation beneath the implant frame.
Sigma-Aldrich
Hydroxyapatite, puriss., meets analytical specification of Ph. Eur., BP, FCC, E341, ≥90% (calculated on glowed substance)
Sigma-Aldrich
Hydroxyapatite, purum p.a., ≥90% (as Ca3(PO4)2, KT)