Merck
  • Inter- and intra-individual variation in urinary biomarker concentrations over a 6-day sampling period. Part 2: personal care product ingredients.
Inter- and intra-individual variation in urinary biomarker concentrations over a 6-day sampling period. Part 2: personal care product ingredients.
Toxicology letters (2014-06-24)
Holger M Koch, Lesa L Aylward, Sean M Hays, Roel Smolders, Rebecca K Moos, John Cocker, Kate Jones, Nicholas Warren, Len Levy, Ruth Bevan
ABSTRACT
An intensive study was conducted to provide data on intra- and inter-individual variation in urinary excretion of a series of ingredients in personal care products (parabens, triclosan, benzophenones) and bisphenol A (BPA, not expected to be an ingredient) in 8 volunteers over 6 days. Exposure diaries recorded use of personal care products with identified target analytes as ingredients. Participants' usual products were replaced with products without the target analytes for 2 of the 6 days. Urine void volumes and times were recorded. Methyl, ethyl, and n-propylparabens, triclosan, benzophenone-3, and BPA were frequently detected (≥70% of samples). Urinary concentrations of the parabens and triclosan were lower on product replacement days. First morning void concentrations correlated moderately to highly with 24-h composite concentrations for all analytes. Intraclass correlation coefficients (ICCs) for spot samples collected on days with usual product use were low for BPA (0.15), moderate for n-propylparaben and methylparaben (0.39 and 0.56, respectively), and high for ethylparaben, benzophenone-3, and triclosan (0.76, 0.81, and 0.934, respectively); ICCs were consistently higher on the basis of cr-adjusted concentrations. Hydration status adjustment methods were assessed by comparing unadjusted and adjusted concentrations to urinary excretion rates (ER, ng/kg-h) for all analytes and samples. Specific gravity-adjusted concentrations correlated slightly better with ER than creatinine-adjusted concentrations. Within-individual variation in biomarker concentrations was highest for methyl and ethylparabens (2 orders of magnitude variation in spot sample concentrations) and lower for the other analytes (1-1.5 orders of magnitude). This dataset provides insight into the design and interpretation of urinary biomonitoring studies for non-persistent chemicals.
Product Description
Supelco
Bisphenol A, ≥99%
Sigma-Aldrich
Bisphenol A, 97%
Sigma-Aldrich
Triclosan, 97.0-103.0% (active substance, GC)
Supelco
Bisphenol A, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Triclosan, Pharmaceutical Secondary Standard; Certified Reference Material