Merck
  • Success with single-agent immunosuppression for multifocal choroidopathies.
Success with single-agent immunosuppression for multifocal choroidopathies.
American journal of ophthalmology (2014-09-10)
Naomi R Goldberg, Theodore Lyu, Erin Moshier, James Godbold, Douglas A Jabs
ABSTRACT
To evaluate the success of single-agent immunosuppression for patients with the posterior uveitides, birdshot chorioretinitis, multifocal choroiditis with panuveitis, and punctate inner choroiditis. Retrospective case series. setting: Tertiary care uveitis practices. population: Patients initiated on immunomodulatory therapy. intervention: Patients were treated with prednisone 1 mg/kg and mycophenolate 2 g daily. Prednisone was tapered after 1 month. Immunosuppression was escalated to mycophenolate 3 g daily, with addition of a second agent, as needed, to achieve treatment success. outcome measure: Treatment success, defined as no disease activity with prednisone dose ≤10 mg daily, at 6, 12, and 24 months. Twenty-seven patients were followed. Mean presentation and 2-year follow-up acuities were 20/41 and 20/42, respectively. For birdshot chorioretinitis, mean (±standard deviation) quantitative Goldmann visual field scores improved from 761 ± 69 degrees (IV/4 isopter) and 496 ± 115 degrees (I/4 isopter) at presentation to 784 ± 57 degrees and 564 ± 125 degrees, respectively. Prednisone was successfully tapered in 95% of patients; mean prednisone doses at 1 and 2 years were 5.3 ± 4.1 and 5.7 ± 4.8 mg/day, respectively. At 2 years, prednisone was discontinued in 11% of patients. Treatment success was achieved in 74% of patients on 1 immunosuppressant, and in an additional 21% of patients on 2 agents, for an overall 95% success rate at 2 years. Posterior uveitides can be treated with 1 agent in most patients, but the data suggest a need to escalate therapy to higher mycophenolate doses, and in one fifth of cases to add a second agent to maintain disease suppression with acceptably low prednisone doses.
Product Description
Supelco
Mycophenolic acid solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
USP
Tacrolimus, United States Pharmacopeia (USP) Reference Standard
Prednisone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
FK-506 monohydrate, ≥98% (HPLC)
Sigma-Aldrich
Prednisone, ≥98%
Sigma-Aldrich
Azathioprine, ≥98%
USP
Mycophenolate mofetil, United States Pharmacopeia (USP) Reference Standard
USP
Prednisone, United States Pharmacopeia (USP) Reference Standard
Azathioprine, European Pharmacopoeia (EP) Reference Standard
Supelco
Prednisone, Pharmaceutical Secondary Standard; Certified Reference Material
Mycophenolate mofetil, European Pharmacopoeia (EP) Reference Standard
USP
Azathioprine, United States Pharmacopeia (USP) Reference Standard
Supelco
Azathioprine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Mycophenolate mofetil, ≥98% (HPLC)
Sigma-Aldrich
(±)-CPP, solid
Sigma-Aldrich
Mycophenolic acid, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Mycophenolic acid, ≥98%
Supelco
Mycophenolic acid, analytical standard
Ciclosporin, European Pharmacopoeia (EP) Reference Standard
Mycophenolate mofetil for peak identification, European Pharmacopoeia (EP) Reference Standard